1,5-Dinitronaphthalene 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13869-14
A13869-22
CAAAA13869-14EA
43.77
CAD
CAAAA13869-14
CAAAA13869-22
1,5-Dinitronaphthalene 97+%
1,5-Dinitronaphthalene
Purity: 97+%
CAS No.: 605-71-0
Molecular Formula: C10H6N2O4
Formula Weight: 218.17
Odor: Odorless
Melting Point: 214-218°
Beilstein Reference: 527184
MDL No.: MFCD00003916
Formula:
C₁₀H₆N₂O₄ Boiling Pt: 390 °C (1013 hPa) Melting Pt: 214…218 °C Density: 1.586 Storage Temperature: Ambient |
MDL Number:
MFCD00003916 CAS Number: 605-71-0 EINECS: 210-095-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...