3-Acetyl-2,5-dichlorothiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2,5-Dichloro-3-thienyl)-1-ethanone
A13867-06
A13867-14
A13867-22
CAAAA13867-06EA
105.23
CAD
CAAAA13867-06
CAAAA13867-14
CAAAA13867-22
3-Acetyl-2,5-dichlorothiophene 98%
3-Acetyl-2,5-dichlorothiophene
Purity: 98%
CAS No.: 36157-40-1
Molecular Formula: C6H4Cl2OS
Formula Weight: 195.07
Flash Point: 110°(230°F)
Melting Point: 37-39°
Boiling Point: 120-122°/4mm
Beilstein Reference: 121288
MDL No.: MFCD00014522
Formula:
C₆H₄Cl₂OS MW: 195.07 g/mol Boiling Pt: 120…122 °C (4 mmHg) Melting Pt: 37…39 °C Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014522 CAS Number: 36157-40-1 EINECS: 252-893-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...