1-Bromo-4-iodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-1-iodobenzene
A13859-30
A13859-09
A13859-18
CAAAAA13859-30EA
1205.54
CAD
CAAAAA13859-30
CAAAA13859-09
CAAAA13859-18
1-Bromo-4-iodobenzene 98%
1-Bromo-4-iodobenzene
Purity: 98%
CAS No.: 589-87-7
Molecular Formula: C6H4BrI
Formula Weight: 282.91
Odor: Recognizable
Melting Point: 89-93°
Boiling Point: 252°
Beilstein Reference: 1904545
MDL No.: MFCD00001051
Formula:
C₆H₄BrI MW: 282.91 g/mol Boiling Pt: 252 °C Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001051 CAS Number: 589-87-7 EINECS: 209-662-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...