(4-Hydroxy-3-nitrophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A13858-14 A13858-06
CAAAAA13858-14EA 1033.32 CAD
CAAAAA13858-14 CAAAA13858-03 CAAAA13858-06
(4-Hydroxy-3-nitrophenyl)acetic acid 99%
(4-Hydroxy-3-nitrophenyl)acetic acid

Purity: 99% CAS No.: 10463-20-4 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 146-148° Beilstein Reference: 2696044

Formula: (C₆H₃(OH)(NO₂))CH₂COOH
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00007122
CAS Number: 10463-20-4
EINECS: 233-948-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR