o-Toluic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13856-0B
A13856-30
CAAAA13856-0BEA
146.36
CAD
CAAAA13856-0B
CAAAA13856-0I
CAAAA13856-30
o-Toluic acid 98+%
o-Toluic acid
Purity: 98+%
CAS No.: 118-90-1
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 148°(298°F)
Odor: Aromatic
Boiling Point: 258-259°
Beilstein Reference: 1072103
Merck Reference: 14,9535
MDL No.: MFCD00002477
Formula:
C₈H₈O₂ Boiling Pt: 258…259 °C Melting Pt: 103…106 °C Density: 1.060 Flash Pt: 148 °C(298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002477 CAS Number: 118-90-1 EINECS: 204-284-9 Merck Index: 14,9535 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...