1-Fluoro-2-nitrobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13852-0B
A13852-18
A13852-30
CAAAA13852-0BEA
971.7
CAD
CAAAA13852-0B
CAAAA13852-18
CAAAA13852-30
1-Fluoro-2-nitrobenzene 99%
1-Fluoro-2-nitrobenzene
Purity: 99%
CAS No.: 1493-27-2
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Flash Point: 94°(201°F)
Physical Form: Liquid
Density: 1.340
Refractive Index: 1.5317
Melting Point: -8°
Boiling Point: 215°
Beilstein Reference: 607262
UN No.: 2810
MDL No.: MFCD00007048
Formula:
C₆H₄FNO₂ Boiling Pt: 215 °C Melting Pt: –8 °C Density: 1.340 Flash Pt: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007048 CAS Number: 1493-27-2 EINECS: 216-088-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...