4-Aminoantipyrine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13846-0E
A13846-22
A13846-36
CAAAAA13846-0EEA
1504.16
CAD
CAAAAA13846-0E
CAAAA13846-22
CAAAA13846-36
4-Aminoantipyrine 97%
4-Aminoantipyrine
Purity: 97%
CAS No.: 83-07-8
Molecular Formula: C11H13N3O
Formula Weight: 203.25
Physical Form: Powder
Appearance: Yellow to pale brown
Melting Point: 106-110°
Beilstein Reference: 181635
Merck Reference: 14,591
MDL No.: MFCD00003145
Formula:
C₁₁H₁₃N₃O Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003145 CAS Number: 83-07-8 EINECS: 201-452-3 Merck Index: 14,591 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...