5-Hydroxysophthalic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13845-0B
A13845-18
A13845-30
CAAAAA13845-0BEA
834.24
CAD
CAAAAA13845-0B
CAAAA13845-18
CAAAA13845-30
5-Hydroxysophthalic acid 98%
5-Hydroxysophthalic acid
Purity: 98%
CAS No.: 618-83-7
Molecular Formula: C8H6O5
Formula Weight: 182.13
Odor: Odorless
Melting Point: ca 300° dec.
Beilstein Reference: 2693561
MDL No.: MFCD00002515
Formula:
C₈H₆O₅ Boiling Pt: 386 °C (1013 hPa) Melting Pt: ∼ 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002515 CAS Number: 618-83-7 EINECS: 210-565-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...