Cyclopropylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13844-09
A13844-18
A13844-30
CAAAA13844-09EA
69.36
CAD
CAAAA13844-09
CAAAA13844-18
CAAAA13844-30
Cyclopropylamine 98+%
Cyclopropylamine
Purity: 98+%
CAS No.: 765-30-0
Molecular Formula: C3H7N
Formula Weight: 57.10
Flash Point: -25°(-13°F)
Refractive Index: 1.4206
Melting Point: -10°
Boiling Point: 49-50°
Beilstein Reference: 741858
UN No.: 2733
MDL No.: MFCD00001301
Formula:
C₃H₇N Boiling Pt: 49…50 °C Melting Pt: –10 °C Density: 0.824 Flash Pt: –25 °C (–13 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001301 CAS Number: 765-30-0 EINECS: 212-142-2 UN: 2733 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...