(±)-1-Phenylethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

A13837-0E A13837-36
CAAAA13837-0EEA 270.18 CAD
CAAAA13837-0E CAAAA13837-36
(±)-1-Phenylethanol 97%
(±)-1-Phenylethanol

Purity: 97% CAS No.: 98-85-1 Molecular Formula: C8H10O Formula Weight: 122.17 Melting Point: 19-20° Boiling Point: 203-205° UN No.: 2937 MDL No.: MFCD00004508

Formula: C₆H₅CH(OH)CH₃
Boiling Pt: 203…205 °C
Melting Pt: 19…20 °C
Density: 1.012
Storage Temperature: Ambient
MDL Number: MFCD00004508
CAS Number: 98-85-1
EINECS: 202-707-1
UN: 2937
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa955c5829772d9b27ccf96aa5e9b66b661e94a5&vid=43f0c1e8ef3c916809a95af92edf59ec33769eab&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR