1,9-Decadiyne 97%

Supplier: Thermo Scientific Chemicals
Warning

A13834-06 A13834-14
CAAAAA13834-22EA 2322.79 CAD
CAAAAA13834-22 CAAAA13834-06 CAAAA13834-14
1,9-Decadiyne 97%
1,9-Decadiyne

Purity: 97% CAS No.: 1720-38-3 Molecular Formula: HC≡C(CH2)6C≡CH Formula Weight: 134.22 Flash Point: 60°(140°F) Refractive Index: 1.4510 Boiling Point: 80-81°/10mm Beilstein Reference: 1734594 UN No.: 3295 MDL No.: MFCD00014926

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 80…81 °C (10 mmHg)
Melting Pt: -10.15
Density: 0.822
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014926
CAS Number: 1720-38-3
EINECS: 217-012-9
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR