Salicylaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Hydroxybenzaldehyde
Precursor to a variey of chelating agents
Purity: 99%
CAS No.: 90-02-8
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 76°(168°F)
Physical Form: Liquid
Density: 1.168
Refractive Index: 1.5730
Sensitivity: Air and Light Sensitive
Odor: Almond-like
Melting Point: -7°
Boiling Point: 197°
Beilstein Reference: 471388
Merck Reference: 14,8326
MDL No.: MFCD00003317
Precursor to a variey of chelating agents
Formula:
C₇H₆O₂ Boiling Pt: 197 °C Melting Pt: –7 °C Density: 1.168 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003317 CAS Number: 90-02-8 EINECS: 201-961-0 UN: 2810 ADR: 6.1,III Merck Index: 14,8326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...