Dansyl chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-(Dimethylamino)naphthalene-1-sulfonyl chloride, 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

A13828-03 A13828-06 A13828-14
CAAAA13828-03EA 76.16 CAD
CAAAA13828-03 CAAAA13828-06 CAAAA13828-14
Dansyl chloride 96%
Dansyl chloride

Used for N terminal amino acid and peptide labeling


Purity: 96+% CAS No.: 605-65-2 Molecular Formula: C12H12ClNO2S Formula Weight: 269.75 Storage Temperature: 2°C to 8°C Odor: Odorless Beilstein Reference: 2217205 Merck Reference: 14,2814 UN No.: 3261 MDL No.: MFCD00003985

Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Melting Pt: 69…75°C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,III
Merck Index: 14,2814

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR