(1-Hexyl)triphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexyltriphenylphosphonium bromide

A13826-14 A13826-22 A13826-36
CAAAA13826-14EA 55.93 CAD
CAAAA13826-14 CAAAA13826-22 CAAAA13826-36
(1-Hexyl)triphenylphosphonium bromide 98%
(1-Hexyl)triphenylphosphonium bromide

Purity: 98% CAS No.: 4762-26-9 Molecular Formula: CH3(CH2)5P(C6H5)3Br Formula Weight: 427.37 Melting Point: 199-202° Beilstein Reference: 3580731

Formula: H₃C(CH₂)₅P(C₆H₅)₃Br
MW: 427.36 g/mol
Melting Pt: 199…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00011860
CAS Number: 4762-26-9
EINECS: 225-301-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39144c363b7dfa31d2778aec18ae322606438bfe&vid=e6acfe673693bc08b35597d39b9c8ca4e35254da&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR