Butyl (S)-(-)-lactate 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-Butyl 2-hydroxypropanoate, n-Butyl L-lactate, (-)-Butyl L-lactate, (S)-(-)-Butyl lactate, Butyl (S)-(-)-2-hydroxypropionate

A13821-0B A13821-0I A13821-30
CAAAA13821-0BEA 175.38 CAD
CAAAA13821-0B CAAAA13821-0I CAAAA13821-30
Butyl (S)-(-)-lactate 97+%
Butyl (S)-(-)-lactate

Purity: 97+% CAS No.: 34451-19-9 Molecular Formula: C7H14O3 Formula Weight: 146.19 Flash Point: 69°(157°F) Refractive Index: 1.4210 Boiling Point: 185-187° MDL No.: MFCD00004519

Formula: C₇H₁₄O₃
Boiling Pt: 185…187 °C
Melting Pt: < 0 °C
Density: 0.984
Flash Pt: 69 °C(157 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004519
CAS Number: 34451-19-9
EINECS: 252-036-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR