2-Bromo-p-toluidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-4-methylaniline, 4-Amino-3-bromotoluene

A13818-09 A13818-18
CAAAAA13818-30EA 1274.41 CAD
CAAAAA13818-30 CAAAA13818-09 CAAAA13818-18
2-Bromo-p-toluidine 99%
2-Bromo-p-toluidine

Purity: 99% CAS No.: 583-68-6 Molecular Formula: C7H8BrN Formula Weight: 186.06 Flash Point: 110°(230°F) Refractive Index: 1.6030 Odor: Characteristic Melting Point: 14-16° Boiling Point: 240-242° Beilstein Reference: 1931711 UN No.: 2810 MDL No.: MFCD00007635

Formula: C₇H₈BrN
Boiling Pt: 240…242 °C
Melting Pt: 14…16 °C
Density: 1.494
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007635
CAS Number: 583-68-6
EINECS: 209-515-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR