Acetonedimethyl acetal 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13810-AE
A13810-0F
A13810-AP
CAAAAA13810-AEEA
43.77
CAD
CAAAAA13810-AE
CAAAA13810-0F
CAAAA13810-AP
Acetonedimethyl acetal 98%
Acetone-dimethyl acetal
Purity: 98%
CAS No.: 77-76-9
Molecular Formula: (CH3O)2C(CH3)2
Formula Weight: 104.15
Flash Point: -11°(12°F)
Physical Form: Liquid
Density: 0.848
Refractive Index: 1.3780
Appearance: Colorless
Odor: Sweet
Melting Point: -47°
Boiling Point: 79-81°
Beilstein Reference: 635678
UN No.: 3271
MDL No.: MFCD00008479
Formula:
C₅H₁₂O₂ Boiling Pt: 79…81 °C Melting Pt: –47 °C Density: 0.848 Flash Pt: –11 °C (12 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008479 CAS Number: 77-76-9 EINECS: 201-056-0 UN: 3271 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...