4-Chlorobenzamide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13809-09
A13809-18
A13809-30
CAAAA13809-09EA
86.9
CAD
CAAAA13809-09
CAAAA13809-18
CAAAA13809-30
4-Chlorobenzamide 98+%
4-Chlorobenzamide
Purity: 98+%
CAS No.: 619-56-7
Molecular Formula: C7H6ClNO
Formula Weight: 155.58
Melting Point: 176-180°
Beilstein Reference: 774622
Formula:
C₇H₆ClNO Boiling Pt: 314 °C (1013 hPa) Melting Pt: 176…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007993 CAS Number: 619-56-7 EINECS: 210-601-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...