2-Fluorobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13800-06
A13800-14
A13800-22
A13800-36
CAAAAA13800-06EA
29.55
CAD
CAAAAA13800-06
CAAAA13800-14
CAAAA13800-22
CAAAA13800-36
2-Fluorobenzaldehyde 97%
2-Fluorobenzaldehyde
Purity: 97%
CAS No.: 446-52-6
Molecular Formula: C7H5FO
Formula Weight: 124.11
Flash Point: 55°(131°F)
Refractive Index: 1.5220
Odor: Odorless
Melting Point: -45°
Boiling Point: 172-174°
Beilstein Reference: 507155
UN No.: 1989
MDL No.: MFCD00003302
Formula:
C₇H₅FO Boiling Pt: 172…174 °C Melting Pt: –45 °C Density: 1.180 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003302 CAS Number: 446-52-6 EINECS: 207-171-2 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...