Caproic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13789-AE
A13789-0F
CAAAAA13789-AEEA
47.97
CAD
CAAAAA13789-AE
CAAAA13789-0F
Caproic acid 98+%
Caproic acid
Purity: 98+%
CAS No.: 142-62-1
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 104°(219°F)
Physical Form: Clear liquid
Density: 0.929
Refractive Index: 1.4165
Appearance: Pale yellow
Melting Point: -4°
Boiling Point: 202-203°
Beilstein Reference: 773837
Merck Reference: 14,1759
UN No.: 2829
MDL No.: MFCD00004421
Formula:
C₆H₁₂O₂ Boiling Pt: 202…203 °C Melting Pt: –4 °C Density: 0.929 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004421 CAS Number: 142-62-1 EINECS: 205-550-7 UN: 2829 ADR: 8,III Merck Index: 14,1759 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...