4-Methoxybenzenesulfonyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13788-14 A13788-22
CAAAA13788-14EA 103.33 CAD
CAAAA13788-14 CAAAA13788-22 CAAAA13788-36
4-Methoxybenzenesulfonyl chloride 98+%
4-Methoxybenzenesulphonyl chloride

Purity: 98+% CAS No.: 98-68-0 Molecular Formula: C7H7ClO3S Formula Weight: 206.65 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 37-42° Boiling Point: 119-122°/0.4mm Beilstein Reference: 609005 UN No.: 3261 MDL No.: MFCD00007446

Formula: C₇H₇ClO₃S
Boiling Pt: 119…122 °C/0.4 mm
Melting Pt: 37…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007446
CAS Number: 98-68-0
EINECS: 202-692-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR