4-(Trichlorosilyl)butyronitrile 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(3-Cyanopropyl)trichlorosilane, Trichloro(3-cyanopropyl)silane
A13787-14
A13787-22
CAAAA13787-14EA
109.02
CAD
CAAAA13787-14
CAAAA13787-22
4-(Trichlorosilyl)butyronitrile 98%
4-(Trichlorosilyl)butyronitrile
Purity: 98%
CAS No.: 1071-27-8
Molecular Formula: C4H6Cl3NSi
Formula Weight: 202.54
Flash Point: 92°(198°F)
Refractive Index: 1.4630
Odor: Pungent
Boiling Point: 237-238°
Beilstein Reference: 906924
UN No.: 2987
MDL No.: MFCD00013832
Formula:
NCH₂CCH₂CH₂SiCl₃ MW: 202.54 g/mol Boiling Pt: 94 Density: 1.28 Flash Pt: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013832 CAS Number: 1071-27-8 EINECS: 213-990-6 UN: 2987 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...