m-Toluic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

A13785-0B A13785-0I A13785-30
CAAAA13785-0BEA 134.62 CAD
CAAAA13785-0B CAAAA13785-0I CAAAA13785-30
m-Toluic acid 99%
m-Toluic acid

Purity: 99% CAS No.: 99-04-7 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 150°(302°F) Melting Point: 108-110° Boiling Point: 263° Beilstein Reference: 970526 Merck Reference: 14,9535 MDL No.: MFCD00002523

Formula: C₈H₈O₂
Boiling Pt: 263 °C
Melting Pt: 108…110 °C
Density: 1.050
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002523
CAS Number: 99-04-7
EINECS: 202-723-9
Merck Index: 14,9535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR