1-Iodohexane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Hexyl iodide

Stabilization: Stabilised with copper

A13784-22 A13784-36
CAAAA13784-22EA 155.95 CAD
CAAAA13784-22 CAAAA13784-36
1-Iodohexane 98% stabilized
1-Iodohexane

Purity: 98% CAS No.: 638-45-9 Molecular Formula: CH3(CH2)5I Formula Weight: 212.08 Flash Point: 61°(141°F) Refractive Index: 1.4920 Melting Point: -75° Boiling Point: 179-181° Beilstein Reference: 505971 MDL No.: MFCD00001102

Formula: C₆H₁₃I
MW: 212.07 g/mol
Boiling Pt: 179…181 °C
Melting Pt: –75 °C
Density: 1.437
Flash Pt: 61 °C(141 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001102
CAS Number: 638-45-9
EINECS: 211-339-0
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR