1-Chloro-2,4-dinitrobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13774-0E
A13774-22
A13774-36
CAAAA13774-0EEA
609.88
CAD
CAAAA13774-0E
CAAAA13774-22
CAAAA13774-36
1-Chloro-2,4-dinitrobenzene 98%
1-Chloro-2,4-dinitrobenzene
Purity: 98%
CAS No.: 97-00-7
Molecular Formula: C6H3ClN2O4
Formula Weight: 202.55
Flash Point: 186°(366°F)
Melting Point: 48-52°
Boiling Point: 314-316°
Beilstein Reference: 613161
Merck Reference: 14,2136
UN No.: 3441
MDL No.: MFCD00007075
Formula:
C₆H₃ClN₂O₄ Boiling Pt: 314…316 °C Melting Pt: 48…52 °C Density: 1.700 Flash Pt: 186 °C (366 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007075 CAS Number: 97-00-7 EINECS: 202-551-4 UN: 3441 ADR: 6.1,II Merck Index: 14,2136 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...