4-Methylbenzylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13768-14
A13768-22
CAAAA13768-14EA
98.28
CAD
CAAAA13768-14
CAAAA13768-22
4-Methylbenzylamine 98%
4-Methylbenzylamine
Purity: 98%
CAS No.: 104-84-7
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 75°(167°F)
Refractive Index: 1.5350
Melting Point: 12-13°
Boiling Point: 196-197°
Beilstein Reference: 956670
UN No.: 2735
MDL No.: MFCD00008123
Formula:
C₈H₁₁N Boiling Pt: 196…197 °C Melting Pt: 12…13 °C Density: 0.955 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008123 CAS Number: 104-84-7 EINECS: 203-243-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...