Phenoxyacetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A13761-18 A13761-30
CAAAA13761-0BEA 1356.45 CAD
CAAAA13761-0B CAAAA13761-18 CAAAA13761-30
Phenoxyacetyl chloride 98%
Phenoxyacetyl chloride

Purity: 98% CAS No.: 701-99-5 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 108°(226°F) Refractive Index: 1.5340 Odor: Irritating Boiling Point: 225-226° Beilstein Reference: 607585 UN No.: 3265 MDL No.: MFCD00000726

Formula: C₈H₇ClO₂
Boiling Pt: 225…226 °C
Melting Pt: 28…30 °C
Density: 1.235
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000726
CAS Number: 701-99-5
EINECS: 211-862-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR