2-Isobutylthiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13759-06
A13759-14
CAAAA13759-06EA
85.64
CAD
CAAAA13759-06
CAAAA13759-14
2-Isobutylthiazole 99%
2-Isobutylthiazole
Purity: 99%
CAS No.: 18640-74-9
Molecular Formula: C7H11NS
Formula Weight: 141.24
Flash Point: 58°(136°F)
Refractive Index: 1.4960
Odor: Characteristic
Boiling Point: 172-173°
Beilstein Reference: 507823
UN No.: 1993
MDL No.: MFCD00005334
Formula:
C₇H₁₁NS MW: 141.24 Boiling Pt: 180 Density: 0.998 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005334 CAS Number: 18640-74-9 EINECS: 242-470-1 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...