4-Aminobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Aminophenyl)(phenyl)methanone, p-Aminobenzophenone

A13757-09 A13757-18
CAAAA13757-09EA 110.13 CAD
CAAAA13757-09 CAAAA13757-18
4-Aminobenzophenone 98%
4-Aminobenzophenone

Purity: 98% CAS No.: 1137-41-3 Molecular Formula: C13H11NO Formula Weight: 197.24 Melting Point: 121-124° Boiling Point: 245-247°/22mm MDL No.: MFCD00007895

Formula: C₁₃H₁₁NO
Boiling Pt: 245…247 °C/22 mm
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3
EINECS: 214-506-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR