2-Chloro-5-nitrobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13754-22 A13754-36
CAAAA13754-22EA 49.45 CAD
CAAAA13754-22 CAAAA13754-36
2-Chloro-5-nitrobenzoic acid 98+%
2-Chloro-5-nitrobenzoic acid

Purity: 98+% CAS No.: 2516-96-3 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Flash Point: 100°(212°F) Odor: Odorless Beilstein Reference: 1877474 MDL No.: MFCD00007294

Formula: C₇H₄ClNO₄
Melting Pt: 166…168 °C
Density: 0.4 g/cm³ (20 °C)
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007294
CAS Number: 2516-96-3
EINECS: 219-739-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR