Benzyl(methyl)amine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13746-AE
A13746-AK
A13746-AU
CAAAAA13746-AEEA
46.29
CAD
CAAAAA13746-AE
CAAAA13746-AK
CAAAA13746-AU
Benzyl(methyl)amine 97%
Benzyl(methyl)amine
Purity: 97%
CAS No.: 103-67-3
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 77°(170°F)
Refractive Index: 1.5230
Odor: Amine-like
pH: 10
Melting Point: -55°
Boiling Point: 184-188°
Beilstein Reference: 606221
UN No.: 2922
MDL No.: MFCD00008289
Formula:
C₆H₅CH₂NHCH₃ Boiling Pt: 184…188 °C Melting Pt: –55 °C Density: 0.940 Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008289 CAS Number: 103-67-3 EINECS: 203-133-4 UN: 2922 ADR: 8,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...