2,3,4-Trihydroxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A13745-06 A13745-14
CAAAA13745-06EA 75.84 CAD
CAAAA13745-06 CAAAA13745-14
2,3,4-Trihydroxybenzaldehyde 98%
2,3,4-Trihydroxybenzaldehyde

Purity: 98% CAS No.: 2144-08-3 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: 159-163° Beilstein Reference: 2328658

Formula: C₇H₆O₄
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 159…163 °C
Density: 1,054 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003325
CAS Number: 2144-08-3
EINECS: 218-404-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR