5-Acetyl-2,4-dimethylthiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13741-06
A13741-14
CAAAA13741-06EA
36.5
CAD
CAAAA13741-06
CAAAA13741-14
5-Acetyl-2,4-dimethylthiazole 99%
5-Acetyl-2,4-dimethylthiazole
Purity: 99%
CAS No.: 38205-60-6
Molecular Formula: C7H9NOS
Formula Weight: 155.22
Flash Point: 104°(219°F)
Refractive Index: 1.5430
Boiling Point: 230°
Beilstein Reference: 120560
MDL No.: MFCD00005331
Formula:
C₇H₉NOS Boiling Pt: 230 °C Density: 1,150 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005331 CAS Number: 38205-60-6 EINECS: 253-826-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...