DL-Tyrosine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Hydroxyphenylalanine, 3-(p-Hidroxifenil)-DL-alanina, H-DL-Tyr-OH
A13740-09
A13740-18
A13740-30
CAAAA13740-09EA
52.46
CAD
CAAAA13740-09
CAAAA13740-18
CAAAA13740-30
DL-Tyrosine 98%
DL-Tyrosine
Purity: 98%
CAS No.: 556-03-6
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Melting Point: ca 325° dec.
Beilstein Reference: 515881
Merck Reference: 14,9839
MDL No.: MFCD00063074
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Boiling Pt: 378 Melting Pt: 285 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063074 CAS Number: 556-03-6 EINECS: 209-113-1 Merck Index: 14,9839 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...