5-Isoxazolecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals

A13739-03 A13739-06
CAAAA13739-03EA 103.33 CAD
CAAAA13739-03 CAAAA13739-06 CAAAA13739-14
5-Isoxazolecarboxylic acid 98%
5-Isoxazolecarboxylic acid

Purity: 98% CAS No.: 21169-71-1 Molecular Formula: C4H3NO3 Formula Weight: 113.07 Melting Point: 141-144° MDL No.: MFCD00156151

Formula: C₄H₃NO₃
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00156151
CAS Number: 21169-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR