2-Aminophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-1-hydroxy benzene, 2-Hydroxy aniline, 2-Hydroxyaniline, o-Aminophenol

A13735-22 A13735-36
CAAAA13735-0EEA 766.57 CAD
CAAAA13735-0E CAAAA13735-22 CAAAA13735-36
2-Aminophenol 99%
2-Aminophenol

Purity: 99% CAS No.: 95-55-6 Molecular Formula: C6H7NO Formula Weight: 109.13 Flash Point: 175°(347°F) Odor: Characteristic Boiling Point: 150°/11mm Beilstein Reference: 606075 Merck Reference: 14,461 UN No.: 2512 MDL No.: MFCD00007690

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 150 °C/11 mm
Melting Pt: 173…177 °C
Density: 1.328
Flash Pt: >175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007690
CAS Number: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 14,461

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR