Ethyl-3,4,5-trihydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Gallic acid ethyl ester, Ethyl 3,4,5-trihydroxybenzoate, Ethyl gallate

A13733-09 A13733-18
CAAAA13733-09EA 80.26 CAD
CAAAA13733-09 CAAAA13733-18
Ethyl-3,4,5-trihydroxybenzoate 98%
Ethyl 3,4,5-trihydroxybenzoate

Purity: 98% CAS No.: 831-61-8 Molecular Formula: C9H10O5 Formula Weight: 198.18 Melting Point: 148-152° Beilstein Reference: 2116014

Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00016430
CAS Number: 831-61-8
EINECS: 212-608-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR