3,5-Dibromosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde

A13729-09 A13729-18
CAAAAA13729-30EA 2275.06 CAD
CAAAAA13729-30 CAAAA13729-09 CAAAA13729-18
3,5-Dibromosalicylaldehyde 98%
3,5-Dibromosalicylaldehyde

Purity: 98% CAS No.: 90-59-5 Molecular Formula: C7H4Br2O2 Formula Weight: 279.93 Melting Point: 81-84° Beilstein Reference: 1424739 Merck Reference: 14,3027 MDL No.: MFCD00003318

Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Boiling Pt: 261
Melting Pt: 86
Storage Temperature: Ambient
MDL Number: MFCD00003318
CAS Number: 90-59-5
EINECS: 202-003-4
Merck Index: 14,3027

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR