4'-Methylacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13721-14 A13721-22 A13721-36
CAAAA13721-14EA 39.03 CAD
CAAAA13721-14 CAAAA13721-22 CAAAA13721-36
4'-Methylacetanilide 98+%
4'-Methylacetanilide

Purity: 98+% CAS No.: 103-89-9 Molecular Formula: C9H11NO Formula Weight: 149.19 Melting Point: 147-150° Boiling Point: 306-308° Merck Reference: 14,74 MDL No.: MFCD00008677

Formula: C₉H₁₁NO
Boiling Pt: 306…308 °C
Melting Pt: 147…150 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008677
CAS Number: 103-89-9
EINECS: 203-155-4
Merck Index: 14,74

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR