Ethyl-4,4,4-trifluoroacetoacetate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13717-09 A13717-18 A13717-30
CAAAA13717-09EA 69.68 CAD
CAAAA13717-09 CAAAA13717-18 CAAAA13717-30
Ethyl-4,4,4-trifluoroacetoacetate 98+%
Ethyl 4,4,4-trifluoroacetoacetate

Purity: 98+% CAS No.: 372-31-6 Molecular Formula: C6H7F3O3 Formula Weight: 184.12 Flash Point: 38°(100°F) Refractive Index: 1.3760 Odor: Odorless Melting Point: -30° Boiling Point: 129-130° Beilstein Reference: 608353 UN No.: 3272 MDL No.: MFCD00000424

Formula: C₆H₇F₃O₃
Boiling Pt: 129…130 °C
Melting Pt: –30 °C
Density: 1.259
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000424
CAS Number: 372-31-6
EINECS: 206-750-7
UN: 1993
ADR: "3",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR