2,3,4,5,6-Pentafluorobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A13713-06 A13713-14
CAAAA13713-06EA 312.84 CAD
CAAAA13713-06 CAAAA13713-14 CAAAA13713-22
2,3,4,5,6-Pentafluorobenzophenone 98%
2,3,4,5,6-Pentafluorobenzophenone

Purity: 98% CAS No.: 1536-23-8 Molecular Formula: C13H5F5O Formula Weight: 272.18 Flash Point: 110°(230°F) Melting Point: 36-39° Boiling Point: 93°/0.2mm Beilstein Reference: 2059098 MDL No.: MFCD00000294

Formula: C₁₃H₅F₅O
MW: 272.17 g/mol
Boiling Pt: 93 °C/0.2 mm
Melting Pt: 36…39 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000294
CAS Number: 1536-23-8
EINECS: 216-258-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR