3-Hydroxy-p-anisic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

A13709-22 A13709-06 A13709-14
CAAAAA13709-22EA 455.04 CAD
CAAAAA13709-22 CAAAA13709-06 CAAAA13709-14
3-Hydroxy-p-anisic acid 97+%
3-Hydroxy-p-anisic acid

Purity: 97+% CAS No.: 645-08-9 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 251-255° Beilstein Reference: 2208365

Formula: C₈H₈O₄
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 251…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00002507
CAS Number: 645-08-9
EINECS: 211-430-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR