Adipic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid

A13705-0B A13705-0G A13705-0I
CAAAA13705-0BEA 63.36 CAD
CAAAA13705-0B CAAAA13705-0G CAAAA13705-0I
Adipic acid 99%
Adipic acid

Purity: 99% CAS No.: 124-04-9 Molecular Formula: HOOC(CH2)4COOH Formula Weight: 146.14 Flash Point: 196°(384°F) Odor: Odorless pH: 2.7 Melting Point: 151-155° Boiling Point: ca 205°/3mm Beilstein Reference: 1209788 Merck Reference: 14,162 MDL No.: MFCD00004420

Formula: HOOC(CH₂)₄COOH
Boiling Pt: ∼ 205 °C/3 mm
Melting Pt: 151…155 °C
Density: 1.360
Flash Pt: 196 °C (384 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 14,162
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ac39e8ebd69527d91cb2d15a469a2a69506785&vid=ef907d9f9fd34903274c42b99d1d1ac1a8db6dce&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR