L(-)-Malic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13702-22
A13702-36
A13702-30
CAAA13702-22EA
121.45
CAD
CAAA13702-22
CAAA13702-36
CAAAA13702-30
L(-)-Malic acid 97%
L(-)-Malic acid
MDL: MFCD00064213
Beilstein: 1723541
Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene
Formula:
C₄H₆O₅ Boiling Pt: 322 °C (1013 hPa) Melting Pt: 100…106 °C Density: 1.60 Flash Pt: 220 °C(428 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064213 CAS Number: 97-67-6 EINECS: 202-601-5 Merck Index: 14,5707 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...