Acetal 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13701-22
A13701-36
CAAAA13701-22EA
55.3
CAD
CAAAA13701-22
CAAAA13701-36
Acetal 99%
Acetal
Purity: 99%
CAS No.: 105-57-7
Molecular Formula: CH3CH(OCH2CH3)2
Formula Weight: 118.18
Storage Temperature: 2°C to 8°C
Flash Point: -21°(-6°F)
Refractive Index: 1.3810
Odor: Pleasant
Melting Point: -100°
Boiling Point: 101-102°
Beilstein Reference: 1098310
Merck Reference: 14,38
UN No.: 1088
MDL No.: MFCD00009243
Formula:
C₆H₁₄O₂ Boiling Pt: 101…102 °C Melting Pt: –100 °C Density: 0.831 Flash Pt: –21 °C (–6 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009243 CAS Number: 105-57-7 EINECS: 203-310-6 UN: 1088 ADR: 3,II Merck Index: 14,38 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...