4-Hydroxybenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
p-Hydroxybenzoic Acid
A13700-0B
A13700-0I
A13700-30
CAAAA13700-0BEA
136.83
CAD
CAAAA13700-0B
CAAAA13700-0I
CAAAA13700-30
4-Hydroxybenzoic acid 99%
4-Hydroxybenzoic acid
Purity: 99%
CAS No.: 99-96-7
Molecular Formula: C7H6O3
Formula Weight: 138.12
Flash Point: 199°(390°F)
Physical Form: Powder
Melting Point: 213-218°
Beilstein Reference: 970950
Merck Reference: 14,4812
MDL No.: MFCD00002547
Formula:
C₇H₆O₃ MW: 138.12 g/mol Boiling Pt: 336 Melting Pt: 213…218 °C Flash Pt: 199 °C (390 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002547 CAS Number: 99-96-7 EINECS: 202-804-9 Merck Index: 14,4812 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...