p-Toluidine 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine

A13698-0B A13698-0I A13698-30
CAAAA13698-0BEA 110.44 CAD
CAAAA13698-0B CAAAA13698-0I CAAAA13698-30
p-Toluidine 99+%
p-Toluidine

Purity: 99+% CAS No.: 106-49-0 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 88°(190°F) Melting Point: 42-46° Boiling Point: 199-202° Beilstein Reference: 471281 Merck Reference: 14,9536 UN No.: 3451 MDL No.: MFCD00007906

Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 199…202 °C
Melting Pt: 42…46 °C
Density: 1.05
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007906
CAS Number: 106-49-0
EINECS: 203-403-1
UN: 3451
ADR: 6.1 II
Merck Index: 14,9536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR