Methyl acetoacetate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-oxobutanoate

A13696-0B A13696-0I A13696-30
CAAAA13696-0BEA 89.43 CAD
CAAAA13696-0B CAAAA13696-0I CAAAA13696-30
Methyl acetoacetate 99%
Methyl acetoacetate

MDL: MFCD00008784 Beilstein: 506727 Soluble in water


Purity: 99% CAS No.: 105-45-3 Molecular Formula: C5H8O3 Formula Weight: 116.12 Flash Point: 70°(158°F) Refractive Index: 1.4185 Odor: Pleasant Melting Point: -80° Boiling Point: 170° Solubility: Soluble in water Beilstein Reference: 506727 Merck Reference: 14,6009 MDL No.: MFCD00008784

Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 170 °C
Melting Pt: –80 °C
Density: 1.076
Flash Pt: 70 °C(158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008784
CAS Number: 105-45-3
EINECS: 203-299-8
Merck Index: 14,6009

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR