1-Chloro-4-fluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13690-14
A13690-22
A13690-36
CAAAA13690-14EA
80.11
CAD
CAAAA13690-14
CAAAA13690-22
CAAAA13690-36
1-Chloro-4-fluorobenzene 98%
1-Chloro-4-fluorobenzene
Purity: 98%
CAS No.: 352-33-0
Molecular Formula: C6H4ClF
Formula Weight: 130.55
Flash Point: 29°(84°F)
Refractive Index: 1.4950
Melting Point: -22°
Boiling Point: 129-130°
Beilstein Reference: 1904542
UN No.: 1993
MDL No.: MFCD00000603
Formula:
C₆H₄ClF Boiling Pt: 129…130 °C Melting Pt: –22 °C Density: 1.228 Flash Pt: 29 °C(84 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000603 CAS Number: 352-33-0 EINECS: 206-521-1 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...