ω,4-Dibromoacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13687-14
A13687-22
CAAAA13687-14EA
115.81
CAD
CAAAA13687-14
CAAAA13687-22
ω,4-Dibromoacetophenone 98+%
ω,4-Dibromoacetophenone
Purity: 98+%
CAS No.: 99-73-0
Molecular Formula: C8H6Br2O
Formula Weight: 277.95
Storage Temperature: 2°C to 8°C
Odor: Pungent
Beilstein Reference: 607604
Merck Reference: 14,1427
UN No.: 3261
MDL No.: MFCD00000200
Formula:
C₈H₆Br₂O Boiling Pt: 310 °C (1013 hPa) Melting Pt: 108…112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000200 CAS Number: 99-73-0 EINECS: 202-783-6 UN: 3261 ADR: 8,III Merck Index: 14,1427 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...